Product Details

SNP ID
rs4704728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:156979597 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTTCTATTAACTTCTAGGTTTT[C/G]CTTTCCCAATTAGTCATTAATCCCT
Phenotype
MIM: 606518
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HAVCR1 PubMed Links

Gene Details

Gene
HAVCR1
Gene Name
hepatitis A virus cellular receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173393.2 Intron NP_001166864.1
NM_001308156.1 Intron NP_001295085.1
NM_012206.3 Intron NP_036338.2
XM_011534515.2 Intron XP_011532817.1
XM_017009339.1 Intron XP_016864828.1
XM_017009340.1 Intron XP_016864829.1

View Full Product Details