Product Details

SNP ID
rs4806366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36858899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCACAGTAACTGGAAGAAAAATC[A/C]AAGTCATGTATCAAGGAAGTAGCGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF345 PubMed Links

Gene Details

Gene
ZNF345
Gene Name
zinc finger protein 345
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242472.1 Intron NP_001229401.1
NM_001242474.1 Intron NP_001229403.1
NM_001242475.1 Intron NP_001229404.1
NM_001242476.1 Intron NP_001229405.1
NM_003419.4 Intron NP_003410.1
XM_011526704.2 Intron XP_011525006.1
XM_017026568.1 Intron XP_016882057.1
XM_017026569.1 Intron XP_016882058.1
XM_017026570.1 Intron XP_016882059.1
XM_017026571.1 Intron XP_016882060.1
XM_017026572.1 Intron XP_016882061.1
XM_017026573.1 Intron XP_016882062.1
Gene
ZNF790
Gene Name
zinc finger protein 790
There are no transcripts associated with this gene.

View Full Product Details