Product Details
- SNP ID
-
rs3136533
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
27
- Location
-
Chr.1:158253270 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTAAAAGAGGAATGTAAATGTGTCC[A/G]GCAAAGACTCATTTATAACAAAACA
- Phenotype
-
MIM: 188370
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD1A
PubMed Links
Gene Details
- Gene
- CD1A
- Gene Name
- CD1a molecule
View Full Product Details