Product Details

SNP ID
rs4350071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:87967662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAGTACTGATGGAGGAGAGCCT[C/G]AGGGTGGCAAGTAAGGAACAGCAGA
Phenotype
MIM: 610076
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDK20 PubMed Links

Gene Details

Gene
CDK20
Gene Name
cyclin dependent kinase 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039803.2 Intron NP_001034892.1
NM_001170639.1 Intron NP_001164110.1
NM_001170640.1 Intron NP_001164111.1
NM_012119.4 Intron NP_036251.2
NM_178432.3 Intron NP_848519.1
XM_017014561.1 Intron XP_016870050.1
XM_017014562.1 Intron XP_016870051.1
XM_017014563.1 Intron XP_016870052.1
XM_017014564.1 Intron XP_016870053.1

View Full Product Details