Product Details

SNP ID
rs4645951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:127736846 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGGAGAAGGCAGAGGGAAAACGG[A/G]AATGGTTTTTAAGACTACCCTTTCG
Phenotype
MIM: 190080
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASC11 PubMed Links

Gene Details

Gene
CASC11
Gene Name
cancer susceptibility candidate 11 (non-protein coding)
There are no transcripts associated with this gene.

Gene
MYC
Gene Name
v-myc avian myelocytomatosis viral oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002467.4 Intron NP_002458.2

View Full Product Details