Product Details

SNP ID
rs1132347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133395741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAAGATGCAGAGCAGCCTGAA[C/G]CTGGTGGACTGTATCATCGAGGTCC
Phenotype
MIM: 615853
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MTG1 PubMed Links

Gene Details

Gene
MTG1
Gene Name
mitochondrial ribosome associated GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138384.2 245 Silent Mutation AAC,AAG N47K NP_612393.2
Gene
PAOX
Gene Name
polyamine oxidase (exo-N4-amino)
There are no transcripts associated with this gene.

View Full Product Details