Product Details

SNP ID
rs4149821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65316436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCAGACGGGGGCAGCCAAAGGT[A/C]AGGATGTAGATGGAGGAGATCTGGG
Phenotype
MIM: 606132
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDC42EP2 PubMed Links

Gene Details

Gene
CDC42EP2
Gene Name
CDC42 effector protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006779.3 Intron NP_006770.1
XM_017017094.1 Intron XP_016872583.1

View Full Product Details