Product Details

SNP ID
rs3179379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184815795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCCGTTGCAGTTCAGCAAGAC[A/G]CTTCCAGTTACTTCCTGCTCTTAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPS8 PubMed Links

Gene Details

Gene
VPS8
Gene Name
VPS8, CORVET complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009921.2 Intron NP_001009921.1
NM_015303.3 Intron NP_056118.2
XM_005247251.3 Intron XP_005247308.1
XM_005247253.4 Intron XP_005247310.1
XM_006713556.3 Intron XP_006713619.1
XM_011512599.2 Intron XP_011510901.1
XM_011512600.2 Intron XP_011510902.1
XM_011512601.2 Intron XP_011510903.1
XM_017006040.1 Intron XP_016861529.1
XM_017006041.1 Intron XP_016861530.1

View Full Product Details