Product Details

SNP ID
rs4902292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64553799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCTGCACACTTTAAGATTTATGC[A/C]CCTTACTACGTGTAAGTTATAACTA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PPP1R36 PubMed Links

Gene Details

Gene
PPP1R36
Gene Name
protein phosphatase 1 regulatory subunit 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172365.2 Intron NP_758953.1

View Full Product Details