Product Details

SNP ID
rs3743463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:96326809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCAGAATTTACTGTTCGCTTTGC[C/T]TCTGCGCTTCATTTTGCAACCTTGT
Phenotype
MIM: 107773
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR1469 PubMed Links

Gene Details

Gene
MIR1469
Gene Name
microRNA 1469
There are no transcripts associated with this gene.

Gene
NR2F2
Gene Name
nuclear receptor subfamily 2 group F member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145155.1 Intron NP_001138627.1
NM_001145156.1 Intron NP_001138628.1
NM_001145157.1 Intron NP_001138629.1
NM_021005.3 Intron NP_066285.1
Gene
NR2F2-AS1
Gene Name
NR2F2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details