Product Details

SNP ID
rs4259128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:107933457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTACGAAGGCATGTAGTGGATGA[A/G]GTGTGAACTGTGACATTTGTTTTTC
Phenotype
MIM: 610670
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP2U1 PubMed Links

Gene Details

Gene
CYP2U1
Gene Name
cytochrome P450 family 2 subfamily U member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183075.2 Intron NP_898898.1
Gene
LOC101929595
Gene Name
uncharacterized LOC101929595
There are no transcripts associated with this gene.

View Full Product Details