Product Details
- SNP ID
-
rs121918793
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:165991549 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTTCCTCTTGTTTTCGTTTTAAA[G/A]TAGTAGTGATTGGCTGATAGGAGAC
- Phenotype
-
MIM: 182389
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LOC102724058
PubMed Links
Gene Details
- Gene
- LOC102724058
- Gene Name
- uncharacterized LOC102724058
There are no transcripts associated with this gene.
- Gene
- SCN1A
- Gene Name
- sodium voltage-gated channel alpha subunit 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001165963.1 |
5931 |
Missense Mutation |
ACT,ATT |
T1909I |
NP_001159435.1 |
NM_001165964.1 |
5931 |
Missense Mutation |
ACT,ATT |
T1881I |
NP_001159436.1 |
NM_001202435.1 |
5931 |
Missense Mutation |
ACT,ATT |
T1909I |
NP_001189364.1 |
NM_006920.4 |
5931 |
Missense Mutation |
ACT,ATT |
T1898I |
NP_008851.3 |
XM_011511602.2 |
5931 |
Missense Mutation |
ACT,ATT |
T1909I |
XP_011509904.1 |
XM_011511604.2 |
5931 |
Missense Mutation |
ACT,ATT |
T1898I |
XP_011509906.1 |
XM_011511605.2 |
5931 |
Missense Mutation |
ACT,ATT |
T1897I |
XP_011509907.1 |
XM_011511606.2 |
5931 |
Missense Mutation |
ACT,ATT |
T1881I |
XP_011509908.1 |
XM_017004644.1 |
5931 |
Missense Mutation |
ACT,ATT |
T1909I |
XP_016860133.1 |
XM_017004645.1 |
5931 |
Missense Mutation |
ACT,ATT |
T1898I |
XP_016860134.1 |
XM_017004646.1 |
5931 |
Missense Mutation |
ACT,ATT |
T1898I |
XP_016860135.1 |
XM_017004647.1 |
5931 |
Missense Mutation |
ACT,ATT |
T1898I |
XP_016860136.1 |
XM_017004648.1 |
5931 |
Missense Mutation |
ACT,ATT |
T1898I |
XP_016860137.1 |
XM_017004649.1 |
5931 |
Missense Mutation |
ACT,ATT |
T1898I |
XP_016860138.1 |
XM_017004650.1 |
5931 |
Missense Mutation |
ACT,ATT |
T1897I |
XP_016860139.1 |
XM_017004651.1 |
5931 |
Missense Mutation |
ACT,ATT |
T1881I |
XP_016860140.1 |
XM_017004652.1 |
5931 |
Missense Mutation |
ACT,ATT |
T1881I |
XP_016860141.1 |
XM_017004653.1 |
5931 |
Missense Mutation |
ACT,ATT |
T1880I |
XP_016860142.1 |
XM_017004654.1 |
5931 |
Missense Mutation |
ACT,ATT |
T1095I |
XP_016860143.1 |
View Full Product Details