Product Details
- SNP ID
-
rs17552296
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:3575333 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCGCGAGATTTGGGTCTCTTCCT[A/C]AGCCGGCGCTCGGCAAGGTAGGTTG
- Phenotype
-
MIM: 603658
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RPS7
PubMed Links
Gene Details
- Gene
- RPS7
- Gene Name
- ribosomal protein S7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001011.3 |
|
Intron |
|
|
NP_001002.1 |
View Full Product Details