Product Details

SNP ID
rs16973434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71698610 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGAGCAACCCATACAAGAAACAG[A/G]CTGAGTACGCTACCAAGGAAGCTGC
Phenotype
MIM: 611066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHLPP2 PubMed Links

Gene Details

Gene
PHLPP2
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289003.1 Intron NP_001275932.1
NM_015020.3 Intron NP_055835.2
Gene
SNORA70D
Gene Name
small nucleolar RNA, H/ACA box 70D
There are no transcripts associated with this gene.

View Full Product Details