Product Details

SNP ID
rs35796589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:21252157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAGTTACAAATAATAGTAAACT[A/G]TCACTTGATACCTACTTGATATTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GREB1L PubMed Links

Gene Details

Gene
GREB1L
Gene Name
growth regulation by estrogen in breast cancer 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142966.1 Intron NP_001136438.1
XM_006722547.2 Intron XP_006722610.1
XM_011526179.2 Intron XP_011524481.1
XM_017025988.1 Intron XP_016881477.1
XM_017025989.1 Intron XP_016881478.1
XM_017025990.1 Intron XP_016881479.1
XM_017025991.1 Intron XP_016881480.1
XM_017025992.1 Intron XP_016881481.1
XM_017025993.1 Intron XP_016881482.1
XM_017025994.1 Intron XP_016881483.1
XM_017025995.1 Intron XP_016881484.1
XM_017025996.1 Intron XP_016881485.1

View Full Product Details