Product Details
- SNP ID
-
rs17025617
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:84821043 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCTATTCAGAGGCCCAGGACCATA[A/G]AGGACACTTGCCACAGGTGGGAGAC
- Phenotype
-
MIM: 603336
MIM: 614912
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DNAH6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78302366] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNAH6
- Gene Name
- dynein axonemal heavy chain 6
There are no transcripts associated with this gene.
- Gene
- TRABD2A
- Gene Name
- TraB domain containing 2A
View Full Product Details