Product Details

SNP ID
rs17131709
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:93183554 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCTTAAATACATCAATGTAAAAT[A/G]TGTGGATTTCATTATGATGTATTGT
Phenotype
MIM: 616876
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC18 PubMed Links

Gene Details

Gene
CCDC18
Gene Name
coiled-coil domain containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306076.1 Intron NP_001293005.1
NM_206886.4 Intron NP_996769.3
XM_011541361.2 Intron XP_011539663.2
XM_011541372.2 Intron XP_011539674.2
XM_017001154.1 Intron XP_016856643.1
XM_017001155.1 Intron XP_016856644.1
XM_017001156.1 Intron XP_016856645.1
XM_017001157.1 Intron XP_016856646.1
XM_017001158.1 Intron XP_016856647.1
XM_017001159.1 Intron XP_016856648.1
XM_017001160.1 Intron XP_016856649.1
XM_017001161.1 Intron XP_016856650.1
XM_017001162.1 Intron XP_016856651.1
XM_017001163.1 Intron XP_016856652.1
XM_017001164.1 Intron XP_016856653.1
XM_017001165.1 Intron XP_016856654.1
XM_017001166.1 Intron XP_016856655.1
XM_017001167.1 Intron XP_016856656.1
XM_017001168.1 Intron XP_016856657.1
XM_017001169.1 Intron XP_016856658.1
XM_017001170.1 Intron XP_016856659.1
XM_017001171.1 Intron XP_016856660.1
Gene
TMED5
Gene Name
transmembrane p24 trafficking protein 5
There are no transcripts associated with this gene.

View Full Product Details