Product Details
- SNP ID
-
rs17209627
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:52792581 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCACTGATGTTCTGACTTCAGTAGA[C/T]AGATGCTCCTCTAACATGTTCCTCC
- Phenotype
-
MIM: 192968
MIM: 605757
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ITGA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80283230] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ITGA1
- Gene Name
- integrin subunit alpha 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181501.1 |
|
Intron |
|
|
NP_852478.1 |
- Gene
- PELO
- Gene Name
- pelota homolog (Drosophila)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015946.4 |
|
Intron |
|
|
NP_057030.3 |
View Full Product Details