Product Details

SNP ID
rs17854049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201380829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAACATCAGGAGGGTTCCGGGAG[G/T]GTCTCAAGCCATACATCTCCTGGAG
Phenotype
MIM: 607334
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRAK2 PubMed Links

Gene Details

Gene
TRAK2
Gene Name
trafficking kinesin protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015049.2 2809 Missense Mutation CAC,CCC H820P NP_055864.2
XM_011511690.1 2809 Missense Mutation CAC,CCC H820P XP_011509992.1
XM_017004772.1 2809 Missense Mutation CAC,CCC H609P XP_016860261.1

View Full Product Details