Product Details

SNP ID
rs17090339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:73688537 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAGGTATACAAAGAGTTGAGGAA[A/C]TGAGAAAGTGTACTCAGGCTAAAAT
Phenotype
MIM: 607531
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLF12 PubMed Links

Gene Details

Gene
KLF12
Gene Name
Kruppel like factor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007249.4 8493 UTR 3 NP_009180.3
XM_005266251.3 8493 UTR 3 XP_005266308.1
XM_011534907.2 8493 UTR 3 XP_011533209.1
XM_011534908.2 8493 Intron XP_011533210.1
XM_011534909.2 8493 UTR 3 XP_011533211.1
XM_011534910.2 8493 UTR 3 XP_011533212.1
XM_011534911.2 8493 UTR 3 XP_011533213.1
XM_011534912.2 8493 UTR 3 XP_011533214.1
XM_017020384.1 8493 UTR 3 XP_016875873.1
XM_017020385.1 8493 UTR 3 XP_016875874.1

View Full Product Details