Product Details

SNP ID
rs17354970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:183100962 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTCCAGACGTGCTAGTTGTTGTC[A/G]TAAACTTCACGCTCCAAGCCAGTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WWC2 PubMed Links

Gene Details

Gene
WWC2
Gene Name
WW and C2 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024949.5 Intron NP_079225.5
XM_011532269.2 Intron XP_011530571.1
Gene
WWC2-AS2
Gene Name
WWC2 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details