Product Details
- SNP ID
-
rs16937191
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:70675012 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATAGTTGTACAGTGGGCAAAAGTTA[C/T]GTATTTCGGCTCGCAGCCAGGTAGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LACTB2
PubMed Links
Gene Details
- Gene
- LACTB2
- Gene Name
- lactamase beta 2
There are no transcripts associated with this gene.
- Gene
- XKR9
- Gene Name
- XK related 9
View Full Product Details