Product Details

SNP ID
rs17110142
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71666686 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTATTTTGTAAATACAAACAAGG[A/T]TTAATTGATTCACTCCTTTTCTACT
Phenotype
MIM: 612531
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
THAP2 PubMed Links

Gene Details

Gene
THAP2
Gene Name
THAP domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031435.3 Intron NP_113623.1
Gene
ZFC3H1
Gene Name
zinc finger C3H1-type containing
There are no transcripts associated with this gene.

View Full Product Details