Product Details

SNP ID
rs16859232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218720199 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTGATGCAGACATGAAACTTGAGA[C/T]GACAAATCTGCAAAATGACTCTTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTLL4 PubMed Links

Gene Details

Gene
TTLL4
Gene Name
tubulin tyrosine ligase like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014640.4 Intron NP_055455.3
XM_006712875.2 Intron XP_006712938.1
XM_017005387.1 Intron XP_016860876.1
XM_017005388.1 Intron XP_016860877.1
XM_017005389.1 Intron XP_016860878.1
XM_017005390.1 Intron XP_016860879.1
XM_017005391.1 Intron XP_016860880.1
XM_017005392.1 Intron XP_016860881.1

View Full Product Details