Product Details

SNP ID
rs34637256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:65895144 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTAAAGCTGATGACATCTTCCAG[A/G]TACTTCTGGCCATATGGTAATTATG
Phenotype
MIM: 608838
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VKORC1L1 PubMed Links

Gene Details

Gene
VKORC1L1
Gene Name
vitamin K epoxide reductase complex subunit 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284342.1 Intron NP_001271271.1
NM_173517.4 Intron NP_775788.2
XM_011515830.2 Intron XP_011514132.1
XM_011515831.2 Intron XP_011514133.1

View Full Product Details