Product Details

SNP ID
rs17623193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68845384 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTATAGAAGACTCTCTAATCTCT[A/T]TGAGTATGTGGTGGAGTACACATAT
Phenotype
MIM: 192090
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CDH1 PubMed Links

Gene Details

Gene
CDH1
Gene Name
cadherin 1
There are no transcripts associated with this gene.

Gene
TANGO6
Gene Name
transport and golgi organization 6 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024562.1 Intron NP_078838.1
XM_011523327.2 Intron XP_011521629.1

View Full Product Details