Product Details

SNP ID
rs17496116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:39950535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCTTTTCCCGGCCGGCTCTGGT[G/T]CTCGGTGTCCCTCCGCCGCCGCTCC
Phenotype
MIM: 603309
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDK13 PubMed Links

Gene Details

Gene
CDK13
Gene Name
cyclin dependent kinase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003718.4 1035 UTR 5 NP_003709.3
NM_031267.3 1035 UTR 5 NP_112557.2
XM_011515597.2 1035 UTR 5 XP_011513899.1
XM_017012750.1 1035 UTR 5 XP_016868239.1
XM_017012751.1 1035 UTR 5 XP_016868240.1

View Full Product Details