Product Details

SNP ID
rs7887165
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:136314022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTCTCACTCTCCACATCTAAT[C/T]CATTGAGTCTTGTTCATGCCACCAC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ADGRG4 PubMed Links

Gene Details

Gene
ADGRG4
Gene Name
adhesion G protein-coupled receptor G4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153834.3 Intron NP_722576.3
XM_011531269.2 Intron XP_011529571.1
XM_011531271.2 Intron XP_011529573.1

View Full Product Details