Product Details
- SNP ID
-
rs17540238
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:89641396 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGTGGAATTGTACCTAGAACAGA[C/T]TGTGAACAGGGGTGCCTGTCAATTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UNQ6494
PubMed Links
Gene Details
- Gene
- UNQ6494
- Gene Name
- uncharacterized LOC100129066
There are no transcripts associated with this gene.
View Full Product Details