Product Details

SNP ID
rs16943328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58974384 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCAGATAACCCTGGAGTTTCAAA[G/T]AACCCTATATCCCTGGGACTTAAAT
Phenotype
MIM: 605073
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PPM1E PubMed Links

Gene Details

Gene
PPM1E
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014906.4 Intron NP_055721.3
XM_005257169.4 Intron XP_005257226.1
XM_011524534.2 Intron XP_011522836.1
XM_017024359.1 Intron XP_016879848.1
Gene
TRIM37
Gene Name
tripartite motif containing 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005207.3 Intron NP_001005207.1
NM_001320987.1 Intron NP_001307916.1
NM_001320988.1 Intron NP_001307917.1
NM_001320989.1 Intron NP_001307918.1
NM_001320990.1 Intron NP_001307919.1
NM_015294.4 Intron NP_056109.1
XM_005257385.1 Intron XP_005257442.1
XM_011524831.1 Intron XP_011523133.1
XM_011524832.2 Intron XP_011523134.1
XM_011524833.1 Intron XP_011523135.1
XM_011524834.1 Intron XP_011523136.1
XM_011524836.1 Intron XP_011523138.1
XM_017024662.1 Intron XP_016880151.1
XM_017024663.1 Intron XP_016880152.1
XM_017024664.1 Intron XP_016880153.1
XM_017024665.1 Intron XP_016880154.1
XM_017024666.1 Intron XP_016880155.1
XM_017024667.1 Intron XP_016880156.1
XM_017024668.1 Intron XP_016880157.1
XM_017024669.1 Intron XP_016880158.1
XM_017024670.1 Intron XP_016880159.1
XM_017024671.1 Intron XP_016880160.1
XM_017024672.1 Intron XP_016880161.1
XM_017024673.1 Intron XP_016880162.1
XM_017024674.1 Intron XP_016880163.1

View Full Product Details