Product Details

SNP ID
rs17118613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:14089678 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTTTGTTAAAAGCAAATACGTCA[C/T]ATGCAGTAGCCATGTAAATACTATA
Phenotype
MIM: 608113
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SGCZ PubMed Links

Gene Details

Gene
SGCZ
Gene Name
sarcoglycan zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322879.1 2364 UTR 3 NP_001309808.1
NM_001322880.1 2364 UTR 3 NP_001309809.1
NM_001322881.1 2364 UTR 3 NP_001309810.1
NM_139167.3 2364 UTR 3 NP_631906.2

View Full Product Details