Product Details

SNP ID
rs16865900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:224379326 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCAGCTGGGTTAGTGCATCTCCAC[C/G]GAACAAAAGCATTCGGTTTTTTTCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM124B PubMed Links

Gene Details

Gene
FAM124B
Gene Name
family with sequence similarity 124 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122779.1 1966 UTR 3 NP_001116251.1
NM_024785.2 1966 UTR 3 NP_079061.2

View Full Product Details