Product Details

SNP ID
rs35834438
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:114397322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTACTCGTATACCAAGTATCCTGC[A/G]CCAATCAATAGCACACAATCATCAA
Phenotype
MIM: 605769
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM33 PubMed Links

Gene Details

Gene
TRIM33
Gene Name
tripartite motif containing 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015906.3 3069 UTR 3 NP_056990.3
NM_033020.2 3069 UTR 3 NP_148980.2
XM_005270936.3 3069 UTR 3 XP_005270993.1
XM_005270937.3 3069 UTR 3 XP_005270994.1
XM_011541568.2 3069 UTR 3 XP_011539870.1
XM_017001452.1 3069 UTR 3 XP_016856941.1
XM_017001453.1 3069 UTR 3 XP_016856942.1
XM_017001454.1 3069 Intron XP_016856943.1

View Full Product Details