Product Details

SNP ID
rs17037754
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:117931919 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGAGTTGAGATTGAATTTTTTTT[G/T]TGTTAGACTAAAACAGCATTTGATA
Phenotype
MIM: 604737
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GDAP2 PubMed Links

Gene Details

Gene
GDAP2
Gene Name
ganglioside induced differentiation associated protein 2
There are no transcripts associated with this gene.

Gene
WDR3
Gene Name
WD repeat domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006784.2 Intron NP_006775.1

View Full Product Details