Product Details
- SNP ID
-
rs17037761
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
17
- Location
-
Chr.1:117932126 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGAACAGATGGGACTGATTTGTTA[G/T]CTCCTTCATTCACCTCTTGTCCCTT
- Phenotype
-
MIM: 604737
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GDAP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116172948] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GDAP2
- Gene Name
- ganglioside induced differentiation associated protein 2
There are no transcripts associated with this gene.
- Gene
- WDR3
- Gene Name
- WD repeat domain 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006784.2 |
|
Intron |
|
|
NP_006775.1 |
View Full Product Details