Product Details

SNP ID
rs17037690
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:117865653 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTATTAGCTTTACTGTCCAACAAA[A/C]GTATCAGATCACATTCTGCAGTTGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GDAP2 PubMed Links

Gene Details

Gene
GDAP2
Gene Name
ganglioside induced differentiation associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135589.2 7152 Intron NP_001129061.1
NM_017686.3 7152 UTR 3 NP_060156.1
XM_005270959.2 7152 UTR 3 XP_005271016.1
XM_011541621.2 7152 Intron XP_011539923.1
XM_011541622.1 7152 Intron XP_011539924.1
XM_011541623.2 7152 UTR 3 XP_011539925.1
XM_011541624.2 7152 Intron XP_011539926.1
XM_017001531.1 7152 UTR 3 XP_016857020.1
XM_017001532.1 7152 UTR 3 XP_016857021.1

View Full Product Details