Product Details

SNP ID
rs16972147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47349722 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCACGAGGCCGAGAGGCGTGGTCG[C/T]CCATGAACTTGGGGAGTGGAGGGAA
Phenotype
MIM: 609949 MIM: 615475
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C5AR2 PubMed Links

Gene Details

Gene
C5AR2
Gene Name
complement component 5a receptor 2
There are no transcripts associated with this gene.

Gene
DHX34
Gene Name
DEAH-box helicase 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014681.5 344 Intron NP_055496.2
XM_005259443.3 344 UTR 5 XP_005259500.1
XM_011527550.2 344 Intron XP_011525852.1
XM_011527551.2 344 Intron XP_011525853.1
XM_017027524.1 344 Intron XP_016883013.1

View Full Product Details