Product Details

SNP ID
rs17853595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:17738107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACATCACGGAGCAAGGACGGCGT[G/T]TGACTGGCCACCTTCTTGGCCAGCA
Phenotype
MIM: 601997
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BCL2L13 PubMed Links

Gene Details

Gene
BCL2L13
Gene Name
BCL2 like 13
There are no transcripts associated with this gene.

Gene
BID
Gene Name
BH3 interacting domain death agonist
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001196.3 589 Missense Mutation CAA,CAC Q162H NP_001187.1
NM_001244567.1 589 Missense Mutation CAA,CAC Q162H NP_001231496.1
NM_001244569.1 589 Missense Mutation CAA,CAC Q66H NP_001231498.1
NM_001244570.1 589 Missense Mutation CAA,CAC Q66H NP_001231499.1
NM_001244572.1 589 Missense Mutation CAA,CAC Q66H NP_001231501.1
NM_197966.2 589 Missense Mutation CAA,CAC Q208H NP_932070.1
NM_197967.2 589 Missense Mutation CAA,CAC Q66H NP_932071.1
XM_017028906.1 589 Missense Mutation CAA,CAC Q162H XP_016884395.1

View Full Product Details