Product Details

SNP ID
rs16973292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71531389 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTCTCTGGTCAGCCCATTGTGTG[C/T]CGTCTAGATCCAGATGTGTCTTGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST4 PubMed Links

Gene Details

Gene
CHST4
Gene Name
carbohydrate sulfotransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166395.1 Intron NP_001159867.1
NM_005769.2 Intron NP_005760.1

View Full Product Details