Product Details

SNP ID
rs17069439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:63828025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATTCTGGCCTATCTGTGCATAC[C/T]ATGTGGAGTGGAACTCATTAGACCA
Phenotype
MIM: 611965
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C3orf49 PubMed Links

Gene Details

Gene
C3orf49
Gene Name
chromosome 3 open reading frame 49
There are no transcripts associated with this gene.

Gene
THOC7
Gene Name
THO complex 7
There are no transcripts associated with this gene.

View Full Product Details