Product Details

SNP ID
rs17473194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:80633029 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGATGTTACACCTAGTAAATTAAC[A/G]GGAGAAAGGTAAGATGAAATCCATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF704 PubMed Links

Gene Details

Gene
ZNF704
Gene Name
zinc finger protein 704
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033723.2 10439 UTR 3 NP_001028895.1
XM_005251279.4 10439 UTR 3 XP_005251336.2
XM_017013725.1 10439 UTR 3 XP_016869214.1

View Full Product Details