Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002646.3 | Intron | NP_002637.3 | |||
XM_005245257.2 | Intron | XP_005245314.1 | |||
XM_005245258.3 | Intron | XP_005245315.1 | |||
XM_011509630.2 | Intron | XP_011507932.1 | |||
XM_011509631.2 | Intron | XP_011507933.1 | |||
XM_011509633.2 | Intron | XP_011507935.1 | |||
XM_017001473.1 | Intron | XP_016856962.1 | |||
XM_017001474.1 | Intron | XP_016856963.1 | |||
XM_017001475.1 | Intron | XP_016856964.1 |