Product Details

SNP ID
rs11558914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:39551636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGAAAACTGCCATCCACAAATA[A/G]ACTACCAAGGTAAATGTAAATACAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMIM14 PubMed Links

Gene Details

Gene
SMIM14
Gene Name
small integral membrane protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317896.1 975 UTR 3 NP_001304825.1
NM_001317897.1 975 UTR 3 NP_001304826.1
NM_174921.2 975 UTR 3 NP_777581.1
XM_011513640.2 975 UTR 3 XP_011511942.1
XM_017007862.1 975 UTR 3 XP_016863351.1
XM_017007863.1 975 UTR 3 XP_016863352.1
Gene
UGDH-AS1
Gene Name
UGDH antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details