Product Details

SNP ID
rs16995383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:39553937 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGTTAGCATGCTACGTATCTTTT[G/T]TGTGTATGTTTTTCGTTTTGATTTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SMIM14 PubMed Links

Gene Details

Gene
SMIM14
Gene Name
small integral membrane protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317896.1 Intron NP_001304825.1
NM_001317897.1 Intron NP_001304826.1
NM_174921.2 Intron NP_777581.1
XM_011513640.2 Intron XP_011511942.1
XM_017007862.1 Intron XP_016863351.1
XM_017007863.1 Intron XP_016863352.1
Gene
UGDH-AS1
Gene Name
UGDH antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details