Product Details

SNP ID
rs35294724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75727180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCATACAGAGTGATGGACCTCAA[C/G]CCGGCTCCCAACCAGTACCAGATGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNM1P35 PubMed Links

Gene Details

Gene
DNM1P35
Gene Name
dynamin 1 pseudogene 35
There are no transcripts associated with this gene.

Gene
ODF3L1
Gene Name
outer dense fiber of sperm tails 3 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175881.3 687 Missense Mutation AAC,AAG N155K NP_787077.1
XM_006720414.3 687 Missense Mutation AAC,AAG N172K XP_006720477.1

View Full Product Details