Product Details

SNP ID
rs34039285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:325455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAACTGCAAGCAGGGTAGGCAA[C/G]AGAGGCCTTAACAACCTCCCTTAGC
Phenotype
MIM: 607621
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COLEC12 PubMed Links

Gene Details

Gene
COLEC12
Gene Name
collectin subfamily member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130386.2 Intron NP_569057.1
XM_011525741.2 Intron XP_011524043.1

View Full Product Details