Product Details

SNP ID
rs16841706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:157414727 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAATTCATAGACTTCAATGGATAT[G/T]AAATAAAACATGGAGTAATTGAAAG
Phenotype
MIM: 604448
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYTIP PubMed Links

Gene Details

Gene
CYTIP
Gene Name
cytohesin 1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004288.4 2293 UTR 3 NP_004279.3
XM_017005386.1 2293 UTR 3 XP_016860875.1

View Full Product Details