Product Details

SNP ID
rs17854022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:105101147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGCCGGTTCAAAAACGTCATCA[C/T]CATCTTCTGCTCCTTCTTCTATCGG
Phenotype
MIM: 604577
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BVES PubMed Links

Gene Details

Gene
BVES
Gene Name
blood vessel epicardial substance
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199563.1 1117 Missense Mutation GAT,GGT D342G NP_001186492.1
NM_007073.4 1117 Missense Mutation GAT,GGT D342G NP_009004.2
NM_147147.3 1117 Missense Mutation GAT,GGT D342G NP_671488.1
XM_011535398.2 1117 Missense Mutation GAT,GGT D342G XP_011533700.1

View Full Product Details