Product Details

SNP ID
rs17345528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101575986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATAATTTGTCTCCTTGCCGTTA[A/C]AAGAAAGTAGTGCTGTTAAGTTTAA
Phenotype
MIM: 615210 MIM: 615423
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PCNP PubMed Links

Gene Details

Gene
PCNP
Gene Name
PEST proteolytic signal containing nuclear protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320395.1 Intron NP_001307324.1
NM_001320397.1 Intron NP_001307326.1
NM_001320398.1 Intron NP_001307327.1
NM_001320399.1 Intron NP_001307328.1
NM_001320400.1 Intron NP_001307329.1
NM_001320401.1 Intron NP_001307330.1
NM_020357.2 Intron NP_065090.1
XM_017006922.1 Intron XP_016862411.1
Gene
TRMT10C
Gene Name
tRNA methyltransferase 10C, mitochondrial RNase P subunit
There are no transcripts associated with this gene.

View Full Product Details