Product Details

SNP ID
rs17583279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:71189412 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAAAATGACCTCTCAGGAGTACA[C/T]TTCTTCCAAAATGAACAGCTAATTT
Phenotype
MIM: 611018
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAIP2B PubMed Links

Gene Details

Gene
PAIP2B
Gene Name
poly(A) binding protein interacting protein 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020459.1 Intron NP_065192.1
XM_005264310.3 Intron XP_005264367.1
XM_005264311.4 Intron XP_005264368.1
XM_011532842.2 Intron XP_011531144.1
XM_011532843.2 Intron XP_011531145.1

View Full Product Details